Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 8 - 9 | 1092 | 111 | |
GenBank mRNA | D17667.1 | - | 7 - 8 | 867 | 111 | |
D38082.1 | - | 7 - 8 | 949 | 111 | ||
S75359.1 | - | 8 - 9 | 1092 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |