Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252682.2 | XP_006252744.1 | 4 - 5 | 630 | 106 | |
XM_008771033.1 | XP_008769255.1 | 4 - 5 | 630 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |