Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004256.1 | NP_001004256.1 | 5 - 6 | 659 | 84 | |
XM_006253801.1 | XP_006253863.1 | 6 - 7 | 843 | 84 | ||
XM_006253802.1 | XP_006253864.1 | 6 - 7 | 721 | 84 | ||
XM_006253803.2 | XP_006253865.1 | 6 - 7 | 715 | 84 | ||
XM_006253804.2 | XP_006253866.1 | 6 - 7 | 666 | 84 | ||
GenBank mRNA | BC078958.1 | - | 5 - 6 | 659 | 7110841 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |