Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004255.1 | NP_001004255.1 | 10 - 11 | 1229 | 139 | |
GenBank mRNA | BC079344.1 | - | 10 - 11 | 1229 | 7107191 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |