Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286936.1 | NP_001273865.1 | 6 - 7 | 735 | 76 | |
XM_006230901.2 | XP_006230963.1 | 6 - 7 | 711 | 76 | ||
XM_008760175.1 | XP_008758397.1 | 6 - 7 | 699 | 76 | ||
GenBank mRNA | BU758664.1 | - | 2 - 3 | 76 | ||
DY546174.1 | - | 4 - 5 | 455 | 76 | ||
FQ227385.1 | - | 6 - 7 | 771 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |