Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053748.1 | NP_446200.1 | 6 - 7 | 814 | 73 | |
XM_006230633.2 | XP_006230695.1 | 7 - 8 | 1001 | 73 | ||
GenBank mRNA | BC107673.1 | - | 7 - 8 | 825 | 7938129 | 73 |
D89340.2 | - | 6 - 7 | 814 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |