Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014172.1 | NP_001014194.1 | 2 - 3 | 343 | 146 | |
XM_008761054.1 | XP_008759276.1 | 2 - 3 | 310 | 146 | ||
XM_008761055.1 | XP_008759277.1 | 2 - 3 | 333 | 146 | ||
GenBank mRNA | BC087665.1 | - | 2 - 3 | 343 | 7317835 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |