Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004232.1 | NP_001004232.1 | 4 - 5 | 1226 | 99 | |
GenBank mRNA | BC079410.1 | - | 4 - 5 | 1226 | 7113273 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |