Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133308.2 | NP_579842.2 | 4 - 5 | 2271 | 101 | |
GenBank mRNA | AF440691.2 | - | 4 - 5 | 2271 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |