Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247236.2 | XP_006247298.1 | 4 - 5 | 1497 | 80 | |
XM_006247237.2 | XP_006247299.1 | 4 - 5 | 1489 | 80 | ||
XM_006247238.2 | XP_006247300.1 | 3 - 4 | 659 | 80 | ||
XM_006247239.2 | XP_006247301.1 | 3 - 4 | 412 | 80 | ||
GenBank mRNA | BC169095.1 | - | 891 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |