Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031344.2 | NP_112634.1 | 3 - 4 | 686 | 67 | |
XM_008760244.1 | XP_008758466.1 | 3 - 4 | 2195 | 67 | ||
GenBank mRNA | AB021980.1 | - | 3 - 4 | 616 | 67 | |
BC081776.1 | - | 3 - 4 | 686 | 7128630 | 67 | |
FQ210893.1 | - | 3 - 4 | 615 | 67 | ||
HQ909027.1 | - | 3 - 4 | 520 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |