Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133535.2 | NP_598219.2 | 6 - 7 | 543 | 139 | |
XM_006231191.2 | XP_006231253.1 | 6 - 7 | 631 | 139 | ||
XM_006231192.2 | XP_006231254.1 | 6 - 7 | 631 | 139 | ||
XM_006231193.2 | XP_006231255.1 | 6 - 7 | 631 | 139 | ||
XM_008760303.1 | XP_008758525.1 | 6 - 7 | 631 | 139 | ||
GenBank mRNA | AF353305.1 | - | 6 - 7 | 599 | 139 | |
BC086376.1 | - | 6 - 7 | 543 | 7135348 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |