Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100666.1 | NP_001094136.1 | 1 - 2 | 77 | 109 | |
GenBank mRNA | CB726734.1 | - | 78 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |