Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY291064.2 | - | 380 | 100 | ||
AY919342.1 | - | 185 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |