Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024888.1 | NP_001020059.1 | 7 - 8 | 1408 | 81 | |
XM_008761144.1 | XP_008759366.1 | 7 - 8 | 1339 | 81 | ||
XM_008761145.1 | XP_008759367.1 | 7 - 8 | 1336 | 81 | ||
GenBank mRNA | BC097428.1 | - | 7 - 8 | 1408 | 7455294 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |