Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138884.1 | NP_620239.1 | 7 - 8 | 896 | 132 | |
XM_006236285.1 | XP_006236347.1 | 7 - 8 | 913 | 132 | ||
GenBank mRNA | D17309.1 | - | 7 - 8 | 896 | 132 | |
FQ211413.1 | - | 7 - 8 | 1065 | 132 | ||
FQ219185.1 | - | 7 - 8 | 912 | 132 | ||
FQ219421.1 | - | 7 - 8 | 856 | 132 | ||
FQ219730.1 | - | 7 - 8 | 857 | 132 | ||
S80431.1 | - | 7 - 8 | 868 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |