Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013079.2 | NP_037211.2 | 5 - 6 | 956 | 67 | |
XM_006236089.2 | XP_006236151.1 | 5 - 6 | 949 | 67 | ||
GenBank mRNA | BC081719.1 | - | 5 - 6 | 956 | 7132312 | 67 |
U07201.1 | - | 5 - 6 | 900 | 67 | ||
U07202.1 | - | 5 - 6 | 900 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |