Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019174.3 | NP_062047.1 | 6 - 7 | 685 | 88 | |
GenBank mRNA | BC097329.1 | - | 6 - 7 | 580 | 7445156 | 88 |
BM387187.1 | - | 2 - 3 | 88 | |||
CB746957.1 | - | 3 - 4 | 246 | 88 | ||
FQ222059.1 | - | 635 | 88 | |||
S68245.1 | - | 6 - 7 | 628 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |