Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044282.1 | NP_001037747.1 | 4 - 5 | 605 | 89 | |
XM_006246126.2 | XP_006246188.1 | 4 - 5 | 755 | 89 | ||
GenBank mRNA | BC085899.1 | - | 4 - 5 | 605 | 7193416 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |