Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113570.1 | NP_001107042.1 | 3 - 4 | 553 | 71 | |
GenBank mRNA | AF391798.1 | - | 3 - 4 | 314 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |