Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109144.1 | NP_001102614.1 | 1 - 2 | 242 | 139 | |
XM_006229118.2 | XP_006229180.1 | 1 - 2 | 141 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |