Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228502.2 | XP_006228564.1 | 4 - 5 | 776 | 123 | |
XM_006228503.2 | XP_006228565.1 | 5 - 6 | 770 | 123 | ||
XM_008759054.1 | XP_008757276.1 | 5 - 6 | 785 | 123 | ||
XM_008759055.1 | XP_008757277.1 | 3 - 4 | 1265 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |