Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cblc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034920.2 | NP_001030092.1 | 10 - 11 | 1509 | 141 | |
GenBank mRNA | BC105776.1 | - | 10 - 11 | 1494 | 7931977 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |