Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198761.1 | NP_942056.1 | 5 - 6 | 1868 | 91 | |
GenBank mRNA | AY382879.1 | - | 5 - 6 | 1868 | 91 | |
FQ228283.1 | - | 2261 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |