Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164060.1 | NP_001157532.1 | 6 - 7 | 1064 | 89 | |
NM_053517.2 | NP_445969.2 | 7 - 8 | 631 | 89 | ||
GenBank mRNA | BC088298.1 | - | 7 - 8 | 631 | 7323045 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |