Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022403.2 | NP_071798.1 | 9 - 10 | 935 | 67 | |
GenBank mRNA | BC089802.1 | - | 9 - 10 | 931 | 7370033 | 67 |
M55167.1 | - | 9 - 10 | 935 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |