Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761118.1 | XP_008759340.1 | 1 - 2 | 187 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |