Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237488.2 | XP_006237550.1 | 1 - 2 | 171 | 71 | |
XM_006237489.2 | XP_006237551.1 | 1 - 2 | 171 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |