Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105828.2 | NP_001099298.2 | 1 - 2 | 90 | 75 | |
GenBank mRNA | BC168730.1 | - | 1 - 2 | 90 | 6887426 | 75 |
FQ213593.1 | - | 1 - 2 | 98 | 75 | ||
FQ217757.1 | - | 1 - 2 | 97 | 75 | ||
FQ224188.1 | - | 1 - 2 | 86 | 75 | ||
FQ228457.1 | - | 1 - 2 | 84 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |