Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053525.1 | NP_445977.1 | 8 - 9 | 1144 | 97 | |
GenBank mRNA | AB055628.1 | - | 8 - 9 | 1144 | 97 | |
BC089107.1 | - | 8 - 9 | 1146 | 7310368 | 97 | |
EF121975.1 | - | 8 - 9 | 1138 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |