Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030023.1 | NP_001025194.1 | 5 - 6 | 684 | 150 | |
GenBank mRNA | AY880882.1 | - | 5 - 6 | 684 | 150 | |
KC795688.1 | - | 6 - 7 | 717 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |