Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024339.1 | NP_001019510.1 | 2 - 3 | 312 | 59 | |
XM_008763254.1 | XP_008761476.1 | 2 - 3 | 449 | 59 | ||
GenBank mRNA | BC079185.1 | - | 2 - 3 | 312 | 7131106 | 59 |
FQ217798.1 | - | 2 - 3 | 368 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |