Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108644.1 | NP_001102114.1 | 4 - 5 | 247 | 147 | |
GenBank mRNA | FQ229611.1 | - | 3 - 4 | 353 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |