Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246362.2 | XP_006246424.1 | 5 - 6 | 875 | 98 | |
XM_006246363.2 | XP_006246425.1 | 5 - 6 | 885 | 98 | ||
XM_006246364.2 | XP_006246426.1 | 5 - 6 | 879 | 98 | ||
XM_008767741.1 | XP_008765963.1 | 5 - 6 | 2699 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |