Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014119.2 | NP_001014141.2 | 5 - 6 | 789 | 79 | |
XM_006247045.2 | XP_006247107.1 | 5 - 6 | 953 | 79 | ||
XM_008768069.1 | XP_008766291.1 | 4 - 5 | 808 | 79 | ||
GenBank mRNA | BC088444.1 | - | 5 - 6 | 783 | 7126427 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |