Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025401.1 | NP_001020572.1 | 13 - 14 | 1267 | 71 | |
GenBank mRNA | BC098031.1 | - | 13 - 14 | 1267 | 7456325 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |