Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053665.1 | NP_446117.1 | 7 - 8 | 2351 | 76 | |
XM_006247067.2 | XP_006247129.1 | 8 - 9 | 2524 | 76 | ||
XM_006247068.1 | XP_006247130.1 | 8 - 9 | 2520 | 76 | ||
XM_008767930.1 | XP_008766152.1 | 8 - 9 | 2579 | 76 | ||
GenBank mRNA | AF068202.1 | - | 7 - 8 | 2351 | 76 | |
BC081753.1 | - | 9 - 10 | 2603 | 76 | ||
BC097298.1 | - | 10 - 11 | 2695 | 76 | ||
BC129090.1 | - | 10 - 11 | 2674 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |