Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039017.1 | NP_001034106.1 | 4 - 5 | 1939 | 133 | |
XM_006246502.2 | XP_006246564.1 | 4 - 5 | 1998 | 133 | ||
XM_006246503.2 | XP_006246565.1 | 2 - 3 | 1797 | 133 | ||
XM_008767821.1 | XP_008766043.1 | 4 - 5 | 1977 | 133 | ||
XM_008767822.1 | XP_008766044.1 | 4 - 5 | 2282 | 133 | ||
XM_008767823.1 | XP_008766045.1 | 4 - 5 | 2285 | 133 | ||
GenBank mRNA | BC099105.1 | - | 4 - 5 | 1939 | 7460369 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |