Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024234.1 | NP_001019405.1 | 3 - 4 | 1125 | 149 | |
GenBank mRNA | BC094520.1 | - | 3 - 4 | 1125 | 7316275 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |