Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135039.1 | NP_001128511.1 | 10 - 11 | 1333 | 84 | |
XM_008768044.1 | XP_008766266.1 | 10 - 11 | 1588 | 84 | ||
GenBank mRNA | CK843896.1 | - | 4 - 5 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |