Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012949.2 | NP_037081.2 | 3 - 4 | 234 | 78 | |
XM_006246599.2 | XP_006246661.1 | 3 - 4 | 305 | 78 | ||
XM_006246600.2 | XP_006246662.1 | 3 - 4 | 285 | 78 | ||
GenBank mRNA | BC083566.1 | - | 3 - 4 | 221 | 7193009 | 78 |
DY321069.1 | - | 3 - 4 | 234 | 78 | ||
FQ214810.1 | - | 2 - 3 | 212 | 78 | ||
FQ214945.1 | - | 3 - 4 | 252 | 78 | ||
FQ214984.1 | - | 3 - 4 | 244 | 78 | ||
FQ215221.1 | - | 2 - 3 | 147 | 78 | ||
FQ215357.1 | - | 3 - 4 | 256 | 78 | ||
FQ215648.1 | - | 3 - 4 | 244 | 78 | ||
FQ215759.1 | - | 3 - 4 | 256 | 78 | ||
FQ215843.1 | - | 2 - 3 | 211 | 78 | ||
FQ215865.1 | - | 3 - 4 | 78 | |||
FQ216150.1 | - | 2 - 3 | 212 | 78 | ||
FQ216382.1 | - | 3 - 4 | 244 | 78 | ||
FQ216386.1 | - | 2 - 3 | 221 | 78 | ||
FQ216660.1 | - | 2 - 3 | 211 | 78 | ||
FQ217095.1 | - | 3 - 4 | 244 | 78 | ||
FQ224938.1 | - | 3 - 4 | 244 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |