Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107057.3 | NP_001100527.1 | 1 - 2 | 365 | 90 | |
GenBank mRNA | BC167045.1 | - | 1 - 2 | 365 | 7112910 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |