Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199489.4 | NP_955783.1 | 1 - 2 | 92 | 88 | |
GenBank mRNA | AY379972.1 | - | 1 - 2 | 95 | 88 | |
BC089762.1 | - | 79 | 7381637 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |