Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252408.2 | XP_006252470.1 | 8 | 6188 | 95 | |
XM_006252409.2 | XP_006252471.1 | 8 | 6183 | 95 | ||
XM_006252410.2 | XP_006252472.1 | 8 | 6254 | 95 | ||
XM_006252413.2 | XP_006252475.1 | 7 | 6036 | 95 | ||
XM_006252415.2 | XP_006252477.1 | 7 | 6033 | 95 | ||
XM_008770870.1 | XP_008769092.1 | 7 | 7990 | 95 | ||
XM_008770871.1 | XP_008769093.1 | 9 | 6200 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |