Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105809.2 | NP_001099279.1 | 7 - 8 | 755 | 105 | |
GenBank mRNA | BC166808.1 | - | 7 - 8 | 755 | 7124384 | 105 |
FQ228687.1 | - | 7 - 8 | 771 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |