Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031651.2 | NP_001026821.1 | 1 - 2 | 509 | 151 | |
GenBank mRNA | BC085794.1 | - | 1 - 2 | 509 | 7109581 | 151 |
BC099074.1 | - | 1 - 2 | 510 | 7109079 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |