Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130415.1 | NP_569099.1 | 4 - 5 | 473 | 72 | |
XM_008767657.1 | XP_008765879.1 | 3 - 4 | 2572 | 72 | ||
XM_008767659.1 | XP_008765881.1 | 5 - 6 | 1155 | 72 | ||
GenBank mRNA | AF361239.1 | - | 4 - 5 | 473 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |