Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012750.1 | NP_036882.1 | 6 - 7 | 1165 | 108 | |
XM_006252238.2 | XP_006252300.1 | 5 - 6 | 1934 | 108 | ||
GenBank mRNA | AF003825.1 | - | 1715 | 108 | ||
AF005226.1 | - | 6 - 7 | 1045 | 108 | ||
U97143.1 | - | 6 - 7 | 1165 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Rattus norvegicus Probe spans exons |