Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100756.1 | NP_001094226.1 | 8 - 9 | 1320 | 58 | |
GenBank mRNA | BC168226.1 | - | 8 - 9 | 1320 | 7371309 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |