Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100756.1 | NP_001094226.1 | 9 - 10 | 1454 | 60 | |
XM_008764464.1 | XP_008762686.1 | 2 - 3 | 2523 | 60 | ||
GenBank mRNA | BC168226.1 | - | 9 - 10 | 1454 | 7371309 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |